Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Arium™ Mini Essential for Production of ASTM Type 1 Ultrapure Water
Compact Arium™ mini laboratory water systems have been designed for Type 1 ultrapure water requirements of 10 liters per day and are ideal for use in preparation of buffers, media and samples, both in life sciences applications and in analytical laboratory procedures.
Supplier: Sartorius H2OMUTUS
Description
- Directly connects to a pretreated water supply line (RO | DI | EDI)
- Reliable: Delivers consistently high water quality for reliable and reproducible results
- Intuitive: Touch-activated color display with direct access to all important dispensing options
- Innovative: saves time-intensive tank cleaning
- Compact: Space-saving, with a width of only 28 cm
Specifications
EU Declaration of Conformity, CSA Declaration of Conformity, QA Certificate | |
10 L | |
Pretreated | |
Type 1 | |
< 5 ppb | |
Lab Water System | |
0 to 6.9 bar; (0 to approx. 100 psi); recommended > 2 bar (>29 psi) | |
20.8 x 11 x 20 in. | |
509 mm | |
100/240 V |
< 100 μS/cm, compensated to 25°C | |
Water pretreated by reverse osmosis, distillation or deionization | |
Up to 1.0 L/min. | |
18.2 Mohm.cm at 25°C | |
<10 ppb | |
Touch activated | |
Arium mini accessories | |
280 mm | |
100/240 VAC 50/60 Hz 2.0 A max | |
Input supply voltage VDC +24 (±10%) Current consumption, max. A 3.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction