Learn More
Epredia™ ArcosSL™ Trays and Storage
Enhance traceability and organization with trays, storage cabinets and cardboard sleeves for the Epredia™ ArcosSL™ slide management system.
Supplier:  Epredia™ A85310012
 View more versions of this product
Catalog No. A85310012
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
Smart and Secure Storage
  • Each Epredia™ ArcosSL™ tray can hold up to 240 slides and features a unique serial number and easy-to-navigate XY coordinate guides. Slides are neatly arranged in the tray, rapidly scanned, and archived in sturdy cabinets, making them easy to identify and locate. Each cabinet can hold up to 40 trays and can be easily configured to fit your facility needs.
  • ArcosSL trays hold slides securely in a shingled manner, allowing maximum visibility to each slide’s tab while maintaining a separation between the slides to avoid damage and cross contamination.  This allows you to optimize tray space and eliminate gaps.
  • ArcosSL cabinets offer flexible storage and easy retrieval of slides in a secure fashion. They can be stacked two units high and safety requirements such as the cabinet base (single or double), bar and fixation ensure your slide’s stability during storage.
  • Cardboard sleeves may be used as an alternative storage option in place of the ArcosSL cabinets. Each sleeve can hold up to four ArcosSL trays.
Specifications
Specifications
Cardboard storage box for four Epredia™ ArcosSL™ trays, Pk. of 25
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Epredia™ ArcosSL™ instrumentation requires the use of ArcosSL trays, sold separately. ArcosSL cabinets are highly recommended for storage.
Single cabinet base is required for single ArcosSL cabinet configuration.
Double cabinet base is required for double ArcosSL cabinet configuration.
ArcosSL cabinet fixation is a safety requirement for ArcosSL cabinets.
ArcosSL cabinet bar is a safety requirement for ArcosSL cabinets.