Learn More
Fine Science Tools Arched Tip Forceps, Stainless Steel, 10 cm
Supplier: Fine Science Tools 1107110
Tip Shape: Straight/ Tips: Arched/ Tip Dimensions: 1.3 x 3 x 0.2mm/ Alloy / Material: Stainless Steel/ Length: 10cm