Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Aquasearcher™ pH Meter AB41PH
SCI_ED

The Aquasearcher™ pH Meter AB41PH is an advanced, research-grade benchtop pH meter with electrode, offering accurate, repeatable results with selectable resolution and an intelligent i-Steward system.

Supplier:  OHAUS™ 30589830/EMD

 View more versions of this product

Catalog No. S05295

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View alternate product


Description

Description

  • Equipped with a large 6.5 in. LCD display and a touch keypad
  • Features a 1000 item memory and password management for GLP documentation
  • Provides connectivity and data output capabilities with RS232 and USB interface
  • i-Steward ensures repeatable and consistent measurement accuracy
  • Intuitive user interface makes changing the parameters, executing setup, and performing calibration as simple as using a smartphone
  • Choose between just the AB41PH meter with electrode holder and a full kit including AB41PH meter, ST410 electrode, STTEMP probe, electrode holder, AS20 magnetic stirrer, and 4 pH buffer mini kits (50 mL)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

±0.3°C
±0.002
USB, RS-232 (PC), RS-232 (Printer)
Auto-stop, Timed, Continuous
-2 to 20
±2000
-10°C to +125°C
LCD
1000 Measurement, 10 Calibration
±0.3
Datalog with Time and Date Stamp
AB41PH Meter, Electrode Holder
Offset
0.001, 0.01, 0.1
Up to 5
LCD, 6.5 in. Segment and Dot Matrix
1000 Sets
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit