Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Aquasearcher™ pH Meter AB33PH PROMO

The Aquasearcher™ pH Meter AB33PH is a highly reliable and user-friendly pH benchtop meter with electrode (ST310) for standard laboratory applications. Designed to measure pH and oxidation-reduction potential with temperature measurements.

Supplier:  OHAUS™ S05296S23

Catalog No. S05296S23

Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative. View substitute products »


Description

Description

  • Equipped with a large 6.5 in. LCD display and a touch keypad
  • Features a 1000 item memory for pH measurements and calibration trail
  • Provides connectivity and data output capabilities with RS232 and USB interface
  • i-Steward monitors the condition of electrodes to ensure repeatable and consistent measurement accuracy
  • Intuitive user interface makes changing the parameters, executing setup, and performing calibration as simple as using a smartphone
  • Auto endpoint mode and auto buffer recognition makes calibration easy
  • Choose between just the AB33PH meter with electrode holder and a full kit including AB33PH meter, ST310 electrode, electrode holder, 4 pH buffer mini kits (50 mL)
Specifications

Specifications

±1
USB, RS-232 (PC), RS-232 (Printer)
Datalog with Time and Date Stamp
Auto-stop, Continuous
-2 to 16
±2000
-5°C to +110°C
LCD
1000 Measurement, Last Calibration
±0.01
±0.5°C
AB33PH Meter, ST310 Electrode, Electrode Holder, 4 pH Buffer Mini Kits (50 mL)
Linear
0.01, 0.1
Up to 3
LCD, 6.5 in. Segment and Dot Matrix
1000 Sets
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit