Learn More
OHAUS™ Aquasearcher™ pH Meter AB23PH PROMO
The Aquasearcher™ pH Meter AB23PH is a simple to use benchtop meter for measuring pH and oxidation-reduction potential (ORP). An intuitive design and auto-buffer recognition make measuring pH straightforward and easy.
Supplier: OHAUS™ S05302S23
Description
- Equipped with a large 5 in. LCD display and six instructional menu buttons
- Auto buffer recognition and 3-point calibration capability ensures a reliable calibration result
- Features a 99 item memory for measurements and last calibration
- Choose between just the AB23PH meter with electrode holder and a full kit including AB23PH meter, ST320 pH electrode, and four pH Buffer Mini Kits (50 mL)
Specifications
±0.5°C | |
±0.01 | |
AB23PH Meter, Electrode Holder, ST320 pH Electrode, 4 pH Buffer Mini Kits (50 mL) | |
Linear | |
0.01 | |
Up to 3 | |
LCD, 5 in. Segment Matrix | |
99 Sets |
±1 | |
Datalog for Measurements and Calibration | |
Auto-stop, Continuous | |
0 to 14 | |
±1999 | |
0°C to 100°C | |
LCD |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.