Learn More
Fluidigm Corp ANTOBODY LABELING KIT 1X-159TB
Supplier:  Fluidigm Corp 201159A
Encompass_Preferred
This Maxpar kit lets you conjugate IgG antibodies with the 159Tb metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.
Catalog No. NC0245971