Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Antipyrine, USP, 98-102%, Spectrum™ Chemical

Click to view available options
Quantity:
100 g
500 g
1 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Antipyrine, USP is a nonsteroidal anti-inflammatory analgesic and antipyretic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 100% |
Ignition Residue | 0.15% |
Loss on Drying | 1% |
Packaging | Poly Pail |
Quantity | 1 kg |
Percent Purity | 98 to 102% |
Grade | USP |