Learn More
TNF Hamster anti-Rat/Mouse, PE, Clone: TN3-19.12, BD
Hamster Monoclonal Antibody
Supplier: BD Biosciences 559503
Description
The TN3-19.12 antibody reacts with rat and mouse tumor-necrosis factor (TNF) proteins (also known as TNF-α) . Moreover, the TN3-19.12 antibody is reported to crossreact with rabbit TNF, but it does not crossreact with mouse lymphotoxin-α (LT-α, also known as TNF-β) nor with human TNF. The immunogen used to generate the TN3-19.12 hybridoma was E. coli-expressed, purified recombinant mouse TNF protein. This monoclonal antibody has been reported to neutralize the bioactivities of mouse, rat and rabbit TNF. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody
development only or reported in the literature.
Intracellular Staining
Specifications
TNF | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant mouse TNF protein | |
0.1 mg | |
Immunology | |
Mouse, Rat | |
IgG1 |
Flow Cytometry | |
TN3-19.12 | |
PE | |
Hamster | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.