Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Rat IgG2a, kappa Isotype Control, Clone: R35-95, BD
Supplier: BD Biosciences 563335
Description
Host Species: Rat
Clone: R35-95
Isotype: IgG2a, kappa
Species Reactivity: Hu, Ms
Immunogen: Mouse Pooled Immunoglobulin
Isotype Control
Specifications
Rat IgG2a, kappa | |
R35-95 | |
Brilliant Violet 786 | |
Affinity Purified | |
RUO | |
Human, Mouse |
Monoclonal | |
0.2mg/mL | |
Rat | |
50 μg | |
Secondary | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction