Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Mouse IgM, κ Isotype Control, Clone: G155-228, BD
Supplier: BD Biosciences 563517
Description
Host Species: Mouse
Clone: G155-228
Isotype: IgM, kappa
Immunogen: TNP-Keyhole Limpet Hemocyanin
Isotype Control
Specifications
Flow Cytometry | |
G155-228 | |
Brilliant Violet 605 | |
Mouse | |
Affinity Purified | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
TNP-Keyhole Limpet Hemocyanin | |
50 μg | |
Isotype Control (Primary) | |
IgM κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction