Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Vibration Nylon Gloves with Grain Leather Tool Grip

Catalog Number p-7113037
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

VEP padding in the palm, thumb webbing, and index finger protects users from cumulative trauma such as bruising from constant tool gripping.

  • Fingerless leather glove with stretchy nylon back
  • Soft grain leather palm cover for longer wear
  • Impact and shock absorbing VEP padding in the palm, between the thumb, and on the index finger
  • Thumb web area is protected from cumulative trauma such as bruising from gripping tools
  • Provides protection from impact, vibration, and abrasion
  • Ideal for use with hand tools and power tools
Color Blue, Yellow
Disposable Reusable
Hand Specific
Material Nylon Lycra
Product Type Anti-impact Gloves
Size X-Small
Sterility Non-sterile
Finger Style Fingerless
For Use With (Application) Construction, DIY Home Improvement
Glove Cuff Style Open
Palm Type Padded
Series 502-20
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit