Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IgG3 κ Isotype Contol, BUV395, Clone: J606, BD
Supplier: BD Biosciences 563814
Description
Host Species: Mouse
Clone: J606
Isotype: IgG3, κ
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
J606 | |
Brilliant Ultraviolet 395 | |
Mouse | |
50 μg | |
Isotype Control (Primary) | |
IgG3 κ |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction