Learn More
IgG, Rat Rat, Polyclonal, Isotype Control, MilliporeSigma™
Rat Isotype Control
Supplier: MilliporeSigma PP68
Description
Rat IgG Polyclonal Antibody control validated for use in ELISA.Specifications
IgG | |
Polyclonal | |
Unconjugated | |
Rat | |
10 mg | |
Maintain for 1 year at 2–8°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap. |
ELISA | |
11.1mg/mL | |
Sterile liquid. Buffer = 0. 01M Sodium Phosphate, 0.25M NaCl, pH 7.6. | |
Ammonium sulfate precipitation | |
Primary | |