Learn More
IgG, Rabbit Rabbit, Polyclonal, Isotype Control, MilliporeSigma™
Rabbit Isotype Control
Supplier:  MilliporeSigma PP64
Catalog No. PP64MI
Add to Cart
Description
Description
Rabbit IgG Polyclonal Antibody validated for use in ELISA.
Specifications
Specifications
IgG
Polyclonal
Unconjugated
Rabbit
25 mg
Maintain at −20°C in undiluted aliquots for up to 12 months. Avoid multiple freeze-thaw cycles.
ELISA
10mg/mL
Frozen nonsterile liquid in a 0.01M Phosphate buffered saline, pH 7.1. Filtered through a 0.45 micron membrane filter.
IgG is purified by cohn fractionation followed by secondary purification with sodium caprylate.
Primary
Purified
Product Suggestions
Product Suggestions