Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG, Goat Goat, Polyclonal, Isotype Control, MilliporeSigma™
Goat Isotype Control
Supplier: MilliporeSigma PP40
Description
Goat IgG Polyclonal Antibody control validated for use in ELISA.Specifications
IgG | |
Polyclonal | |
Unconjugated | |
Goat | |
Primary | |
Purified |
ELISA | |
4.59mg/mL | |
Purified immunoglobulin. Liquid filtered through 0.45 μm Swinex, no preservatives. | |
50 mg | |
Store −20°C for up to 12 months. Avoid repeated freeze thaw cycle. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction