Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-γ Mouse anti-Pig, PE, Clone: P2G10, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 559812
Description
The P2G10 antibody reacts with porcine interferon-γ (IFN-γ). The immunogen used to generate the P2G10 hybridoma was recombinant pig IFN-γ.
Intracellular Staining
Specifications
IFN-γ | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant pig IFN-γ | |
0.1 mg | |
Immunology | |
Porcine | |
IgG1 κ |
Flow Cytometry | |
P2G10 | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction