Learn More
CD49b Hamster anti-Mouse, PE, Clone: HMA2, BD
Hamster Monoclonal Antibody
Supplier: BD Biosciences 558759
Description
The HM α 2 antibody reacts with integrin α 2 chain (CD49b), the 150-kDa transmembrane glycoprotein that non-covalently associates with the integrin β 1 subunit (CD29) to form the integrin α 2β 1 complex known as VLA-2. VLA-2, a receptor for collagen and laminin, is expressed on some splenic CD4+ T lymphocytes and NK-T cells, intestinal intraepithelial and lamina propria lymphocytes, splenic NK cells, epithelial cells, and platelets; but it is not on thymocytes or Peyer’s-patch or lymphnode lymphocytes. The expression of VLA-2 is upregulated on lymphocytes in response to mitogens. The HM α 2 antibody has been reported to partially block the interaction of T-cell blasts, but not NK cells, with collagen. Purified HM α 2 mAb blocks the staining of splenic NK cells by the anti-CD49b/Pan-NK Cells mAb DX5 (Cat. No. 553858, for the PE conjugate). Therefore, mAb HM α 2 may be used like the DX5 mAb for identification of NK cells.
Specifications
CD49b | |
Monoclonal | |
0.2mg/mL | |
Integrin α2 chain | |
Hamster | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
HMA2 | |
PE | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse colon carcinoma cell line Colon26 | |
0.1 mg | |
Immunology | |
Mouse | |
IgG1 κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.