Learn More
CD28 Mouse anti-Human, APC, Clone: CD28.2, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 559770
Description
Recognizes a 44kDa homodimeric transmembrane glycoprotein present on most mature T cells, thymocytes and plasma cells. CD28 is the ligand for CD80 and plays a very important role in T cell-B cell interaction. It has been suggested that CD28 initiates and regulates a separate and distinct signal transduction pathway from those stimulated by the TCR complex. Additionally, it has been reported that CD28 antibody clones vary in their ability to stimulate T cells to produce IL-2 and increase intracellular Ca2+ concentration. This finding suggests the existence of functionally distinct subregions on the CD28 molecule. CD28.2 has been demonstrated to bind to the same molecule as clone L293, another CD28 mAb, and has been reported to induce Ca2+ influx in Jurkat T cells.
Specifications
CD28 | |
Monoclonal | |
APC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human CD28 Transfected Cell Line | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
CD28.2 | |
CD28 antigen; T44; Tp44; TP44 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.