Learn More
CD227 Mouse anti-Human, FITC, Clone: HMPV, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 559774
Description
The HMPV monoclonal antibody specifically binds to CD227 which is also known as Mucin-1 (MUC1). A major form of CD227 is expressed as a type I transmembrane glycoprotein. CD227 belongs to the epithelial mucin family whose members are heavily O-glycosylated and characterized by high molecular weight, and an amino acid composition rich in serine, threonine, proline, and glycine. CD227 is variably expressed on the surfaces of normal and malignant glandular and ductal epithelial cells, and some hematopoietic cell lineages including subsets of T cells, B cells, monocytes and dendritic cells. Soluble forms of CD227 may arise by shedding from the cell surface or by secretion of forms derived from alternative RNA splicing. The HMPV antibody binds to the core peptide of the MUC1 protein. The core protein contains a domain of 20 amino-acid tandem repeats which function as multiple epitopes for this monoclonal antibody. Incomplete glycosylation of some tumor-associated mucins may lead to variable unmasking of the multiple peptide epitopes leading to the observed differences in immunostaining intensities between cells from normal and malignant tissues. CD227 plays roles in the provision of protective barrier function, the regulation of cellular adhesion, and the transduction of multiple signal pathways.
Specifications
CD227 | |
Monoclonal | |
FITC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Milk Fat Globule Membranes | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
HMPV | |
MUC1; CA 15-3; DF3; EMA; Episialin; H23; H23AG; KL-6; MAM6; PEM; PEMT; PUM | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.