Learn More
CD195 Rat anti-Mouse, PE, Clone: C34-3448, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 559923
Description
The C34-3448 antibody reacts with mouse chemokine receptor CD195 (also known as CCR5), a member of the β - chemokine receptor family. CD195 regulates lymphocyte chemotaxis activation and transendothelial migration during inflammation. It signals a response to at least three chemokines: RANTES and macrophage inflammatory protein-1 (MIP-1) α and β . In the mouse system, the gene for CD195 has been mapped to chromosome 9. CD195 mRNA is expressed in heart, spleen and liver tissues, and by macrophages and T-lymphocytes. The immunogen used to generate the C34- 3448 hybridoma was a KLH-conjugated peptide consisting of amino acids 9-30 of mouse CD195 (CCR5).
Specifications
CD195 | |
Monoclonal | |
0.2mg/mL | |
CCR5 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2c κ |
Flow Cytometry | |
C34-3448 | |
PE | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse CCR5 aa. 9-30 | |
0.2 mg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.