Learn More
CD13 Rat anti-Mouse, PE, Clone: R3-242, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 558745
Description
The R3-242 monoclonal antibody specifically binds to aminopeptidase N (APN), which has been reported to be the mouse CD13 homologue. APN is expressed in a variety of mammalian tissues, including intestinal and renal epithelial brush border, endothelium, liver, lung, and brain. In lymphoid tissues, R3-242 mAb is reported to detect APN on monocytes, macrophages, mast cells, dendritic cells, and B lymphocytes, but not on T cells, and thymocytes. However, the reported reactivity with B cells may be non-specific, via binding to the Fcγ receptors (CD16/CD32). This observation is in agreement with reports using different antibodies to mouse APN. APN may play a role in antigen presentation in the immune system. R3-242 mAb does not inhibit the enzymatic activity of APN.
Specifications
CD13 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Crude Aminopeptidase N Preparation from BALB/c Mouse Intestinal Epithelium | |
0.1 mg | |
Immunology | |
Mouse | |
IgG1 |
Flow Cytometry | |
R3-242 | |
PE | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.