Learn More
Ansell™ Sawyer-Tower™ 66-672 Series Bib Overalls
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Description
- Features comfortable Gore™ Chemical Splash Fabric
- Chemical splash protection
- Breathable Gore-Tex fabric keeps workers cool in hot situations and warm in cold situations
- Lightweight, flexible, snag- and tear-resistant, stain-repellent and launderable
- CPC Nomex provides fire-resistant chemical splash protection
- Special sizes may be available upon request
- Flame resistance
- HRC rating: 2
Specifications
Color | Blue |
Material | CPC Nomex Trilaminate |
Size | Medium |
Arc Flash Rating | 2 |