Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell Beard Covers
Click to view available options
Regular
Description
- Lightweight polypropylene
- White
- Packaged 100/pack
Specifications
Type | General Purpose Beard Cover |
Size | Regular |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
For Use With (Application) | Economical protection for light to medium duty use |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction