Learn More
TARGETMOL CHEMICALS INC ANGIOTENSIN II HUMAN ACE 25MG
Supplier:  TARGETMOL CHEMICALS INC T8560
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Angiotensin II human acetate (DRVYIHPF acetate) is a vasoconstrictor that mainly acts on the AT1 receptor. purity: 99%
Catalog No. 50-257-6234