Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troemner™ Analytical Precision UltraClass Weight Set, No Certificate
Combine high precision with the advantage of two-piece construction with the most precise two-piece weights available with weight tolerances that equal ANSI/ASTM E617 Class 0 or OIML R 111 Class E2 tolerances.
$1772.00 - $3453.00
Specifications
Certifications/Compliance | No Certificate |
---|---|
Classification | UltraClass |
Case Material | Polycarbonate |
Length (Metric) | 20.32 cm |
Height (Metric) Case | 8.89 cm |
Catalog Number | Mfr. No. | Weight (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Weight (Metric) | Price | Quantity & Availability | |||||
02-217-055
|
Troemner™
30390076 |
100 mg |
Each for $1,811.00
|
|
|||||
02-217-052
|
Troemner™
30390079 |
1 to 200 g |
Each for $1,772.00
|
|
|||||
02-217-049
|
Troemner™
30390009 |
(2) 200 g to 1 g |
Each for $2,023.00
|
|
|||||
02-217-045
|
Troemner™
30390081 |
500 to 1 g |
Each for $3,453.00
|
|
|||||
Description
- Used in extremely stable environments
- 1 to 500 mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
- Weights 1 g and above are of two-piece construction
- Weight sets 1 mg-500 g are provided in a high-quality polycarbonate set case
Specifications
No Certificate | |
Polycarbonate | |
8.89 cm | |
Calibration of high-precision analytical balances with readability as low as 0.01 mg |
UltraClass | |
20.32 cm | |
Reference standard calibration |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title