Learn More
Analyte Mix, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep 8141AB
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Aspon | 3244-90-4 | 200μg/mL |
Azinphos-ethyl | 2642-71-9 | 200μg/mL |
Carbophenothion | 786-19-6 | 200μg/mL |
Chlorfenvinfos | 470-90-6 | 200μg/mL |
Chlorpyrifos-methyl | 5598-13-0 | 200μg/mL |
Crotoxyphos | 7700-17-6 | 200μg/mL |
Dichlorfenthion | 97-17-6 | 200μg/mL |
Dicrotophos | 141-66-2 | 200μg/mL |
Dimethoate | 60-51-5 | 200μg/mL |
Dioxathion | 78-34-2 | 200μg/mL |
Dyfonate (Fonofos) | 944-22-9 | 200μg/mL |
Dylox | 52-68-6 | 200μg/mL |
EPN | 2104-64-5 | 200μg/mL |
Ethion | 563-12-2 | 200μg/mL |
Ethyl parathion | 56-38-2 | 200μg/mL |
Fenitrothion | 122-14-5 | 200μg/mL |
Imidan | 732-11-6 | 200μg/mL |
Leptophos | 21609-90-5 | 200μg/mL |
Malathion | 121-75-5 | 200μg/mL |
Monocrotophos | 6923-22-4 | 200μg/mL |
Phosphamidon(33.4% E; 64.6% Z) | 13171-21-6 | 200μg/mL |
TEPP | 107-49-3 | 200μg/mL |
Terbufos | 13071-79-9 | 200μg/mL |
Tetraethyldithio Pyrophosphate/Sulfotepp | 3689-24-5 | 200μg/mL |
Zinophos | 297-97-2 | 200μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Analyte Mix | |
2mL amber prescored glass ampule | |
Hexane:Acetone | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
SW-846 Method 8141 | |
For US EPA Method 8141 | |
1 mL | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Freezer
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product intended for laboratory use only