Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amylose, Spectrum™ Chemical
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Amylose , a helical polymer, is made of α-D-glucose units and is important in plant energy storage; it can be used in industrial and food based contexts as a water binder, thickener, emulsion stabilizer, and gelling agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
CAS Max % | 100% |
Quantity | 1 g |
Grade | Reagent |
Packaging | Amber Glass Bottle |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction