Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Sulfate, TraceSELECT™, Honeywell™

Catalog Number 6001885
Click to view available options
Quantity:
100 g
7783-20-2
H8N2O4S
132.13
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
6097028
CHEBI:62946
diazanium;sulfate
N.N.OS(O)(=O)=O
7783-20-2
H8N2O4S
132.13
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
6097028
CHEBI:62946
diazanium;sulfate
N.N.OS(O)(=O)=O

TraceSELECT™, for trace analysis,≥99.9999% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY
Quantity 100 g
Linear Formula (NH4)2SO4
UN Number NONH for all modes of transport
Solubility Information Completely Soluble in water , 132g/L at 20°C
Formula Weight 132.14g/mol
Percent Purity ≥99.99% (metals basis)
Grade TraceSELECT™
Density 1.77g/mL at 25°C
Chemical Name or Material Ammonium Sulfate
EINECSNumber 231-984-1
RTECSNumber BS4500000
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit