Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Bromide (Powder/Certified ACS), Fisher Chemical

Catalog Number A650500
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
12124-97-9
BrH4N
97.94
MFCD00011419
SWLVFNYSXGMGBS-UHFFFAOYSA-N
ammonium bromide
[NH4+].[Br-]
12124-97-9
BrH4N
97.94
MFCD00011419
SWLVFNYSXGMGBS-UHFFFAOYSA-N
ammonium bromide
[NH4+].[Br-]
Boiling Point 235°C
Melting Point 452°C
Color White
pH 4.5 to 5.5
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range ≥99.0 %
Formula Weight 97.94
Percent Purity ≥99.0%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Chemical Name or Material Ammonium Bromide
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit