Learn More
Promega Amino Acid Mixture Minus Cysteine
Amino Acid Mixture, Complete, is an aqueous solution containing 1mM each of the 20 essential amino acids.
Supplier:  Promega L4471
Catalog No. PRL4471
Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative.
Description
Description
  • 1mM Solution of 20 Essential Amino Acids or Combinations Thereof
  • Amino acid solution for use with Flexi(R), TnT(R) and Rabbit Reticulocyte Lysates, Wheat Germ Extract and E. coli S30 Systems
  • Choose from mix of 20 amino acids or special mixtures indicated
  • Supplied at a concentration 1mM
Specifications
Specifications
1mM of each Amino Acid
<-65°C
175 μL