Learn More
AccuTec Blades™ Gem Scientific, 3 Facet, Aluminum Back, .009' Carbon, 100 Blade Bulk Dispenser
Precise, sharp and safe blade suitable for a variety of industrial applications
Supplier: AccuTec Blades™ 6203250000
Description
- Three-facet blade with aluminum back
- Safe dispensing — simply push button to release/eject blade; blade pops up with backing first
- See-through cover reveals number of remaining blades
- Secure, built-in used blade storage
- 100 blades/dispenser
Specifications
Carbon Steel | |
Razor Blade |