Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Educational Products Student Distilling Apparatus
SCI_ED

Quality distillation equipment for chemistry labs.

Supplier:  American Educational Products 74002/PROMO

Catalog No. S50812PR

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. View substitute products »


Includes:
  • 11.8 in. (300mm) Liebig condenser
  • Kjeldahl distilling column with 50mm diameter bulb
  • 500mL boiling flask
  • Tripod base, rod and clamp
  • Iron tripod and wire gauze
  • 500mL Erlenmeyer flask for distillate
  • Comprehensive instruction manual

Description

Description

No two liquids have exactly the same boiling point, therefore, it is possible to separate any mixture of two liquids by raising the temperature of the mixture to a point where one of the liquids will boil but the other remains liquid.

  • Dimensions: 24 × 6.25 × 7.75 in. (60.9 × 15.9 × 19.7cm)
  • Weight: 8 lb. (0.45kg)

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit