Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Educational Products Flint Glass Wide-Mouth Jars
SCI_ED

Click to view available options
Capacity (Metric):
237 mL
473 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Lighter and more economical.

Wide-mouth, machine-made, nonporous flint glass bottles.

  • Lighter than handmade bottles and less expensive
  • Suitable for gas generation and collection
  • Easily cleaned and autoclavable

Product Type Jar
Capacity 16 oz., 473 mL
Capacity (Metric) 473 mL
Capacity (English) 16 oz.
For Use With (Application) Gas Generation and Collection
Material Glass
Closure Included No
Mouth Wide
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit