Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amerex™ Wheeled Fire Extinguishers: Multipurpose ABC Dry Chemical
Wheeled, dry chemical fire extinguisher
$9180.00 - $14790.00
Specifications
Length (English) Hose | 50 ft. |
---|---|
Length (Metric) Hose | 15 m |
Range | 30 to 40 ft. |
Reagent Type | ABC dry chemical |
Temperature Range (English) | -65° to +120°F |
Catalog Number | Mfr. No. | Capacity (English) | Product Type | Discharge Time | UL Rating | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Product Type | Discharge Time | UL Rating | Price | Quantity & Availability | ||
19-860-542
|
Amerex™
11483 |
125 lb. | Semi-pneumatic Wheeled Fire Extinguisher | 52 sec. | 40A:240B:C |
Each for $9,180.00
|
|
19-860-544
|
Amerex™
15501 |
125 lb. | Semi-pneumatic Wheeled Fire Extinguisher | 48 sec. | 40A:240B:C |
Each for $10,560.00
|
|
19-860-550
|
Amerex™
11677 |
300 lb. | Steel-wheeled Fire Extinguisher | 60 sec. | 40A:320B:C |
Each for $14,790.00
|
|
19-860-547
|
Amerex™
10701 |
125 lb. | Steel-wheeled Fire Extinguisher | 48 sec. | 40A:240B:C |
Each for $13,860.00
|
|
Description
- Semi-pneumatic wheels
- Coast Guard approved
- Contains a specially fluidized and siliconized mono ammonium phosphate chemical
- Chemically insulates Class A fires by melting at approximately 350°F, coating the surface to which it is applied
- Smothers and breaks the chain reaction of Class B fires and will not conduct electricity
Specifications
50 ft. | |
30 to 40 ft. | |
-65° to +120°F |
15 m | |
ABC dry chemical | |
-54°C to +49°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title