Learn More
Amerex™ Water Mist Fire Extinguishers
Ultimate extinguisher for Class A fires where a potential Class C (electrical) hazard exists
$586.00 - $616.00
Specifications
Product Type | Fire Extinguisher |
---|---|
Material | Stainless Steel |
Includes | Non Magnetic wall bracket |
Length (English) | 7 in. |
Length (Metric) | 17.78 cm |
Catalog Number | Mfr. No. | Discharge Time | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Discharge Time | Model | Price | Quantity & Availability | ||||
19-860-525
|
Amerex™
16374 |
72 sec. | B270NM |
Each for $586.00
|
|
19-860-526
|
Amerex™
16376 |
80 sec. | B272NM |
Each for $616.00
|
|
Description
The fine spray from the unique misting nozzle provides safety from electrical shock, greatly enhances the cooling and soaking characteristics of the agent and reduces scattering of burning materials.
- Stored pressure design
- Stainless steel cylinders
- Durable high gloss polyester powder paint
- All metal valve construction
- Exclusive crevice free, butt welded cylinder
- Temperature range: +40 to 120°F
- Nonmagnetic construction tested by an independent agency for use in MRI facilities to 11.7 Tesla (including wall hanger bracket)
- Non-toxic
- De-ionized water extinguishing agent (sold separately)
- Wand provides greatest operator safety
- Large loop stainless steel pull pin
- Maximum visibility during discharge
- No electrical conductivity back to the operator
- No thermal or static shock
- Bar coded and bi-lingual labels
Warranty and Services
5 Year.
Specifications
Fire Extinguisher | |
Non Magnetic wall bracket | |
17.78 cm | |
Deionized Water | |
4.4°C to 48.8°C | |
11 in. |
Stainless Steel | |
7 in. | |
10 to 12 ft. | |
40° to 120°F | |
2A:C | |
27.94 cm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.