Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
American Educational Products Bowles Stethoscope
Exceptional listening ability!
Supplier: American Educational Products 71872
Alert:
This item is intended for educational purposes only.
Description
Designed for many basic diagnostic uses.
- Economical
- Light and adjustable
- Sensitive chest piece
Order Info
Dimensions: 7.75 x 4 x 1 in. , Weight: 11 lb. (5kg)
Specifications
Bowles Stethoscope | |
Sensitive chest piece | |
49.89g |
For many basic diagnostic uses | |
0.11 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction