Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bal Supply Latex Pipette Bulbs
Small dropper bulbs made of natural latex.
$22.25 - $32.75
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | Amber |
Disposable | Single-use |
For Use With | Pasteur Pipette, Small Pipette, Medicine Dropper |
Material | Latex |
Description
For use with Pasteur Pipettes, Small Pipettes and Medicine Droppers.Specifications
Not Autoclavable | |
Single-use | |
Latex | |
Non-sterile |
Amber | |
Pasteur Pipette, Small Pipette, Medicine Dropper | |
1 | |
Amber Latex Bulb |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title