Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum, 6 x 6 x 0.0625 in., Sheet, Spectrum™ Chemical
SDP

Click to view available options
Quantity: 100 g
7429-90-5
7429-90-5
CAS Max % 1
Quantity 100 g
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.