Learn More
Integrated DNA Technologies Inc Alt-R CRISPR tracrRNA, 5 nmol
Supplier: Integrated DNA Technologies Inc 1072532
Universal 67mer tracrRNA that contains proprietary chemical modifications conferring increased nuclease resistance. Hybridizes to crRNA to activate the Cas9 enzyme.