Learn More
Integrated DNA Technologies Inc Alt-R A.s. Cas12a (Cpf1) V3, 100 µg
Supplier:  Integrated DNA Technologies Inc 1081068
Encompass
Recombinant Acidaminococcus sp. BV3L6 Cas12a (Cpf1) nuclease, purified from an E. coli strain expressing Cas12a. Contains nuclear localization sequence (NLS) and C-terminal 6-His tag. Provided in solution at 10 µg/µL. 100 µg of Cas12a nuclease = 633 pmol.
Catalog No. NC1548675
This product needs to be quoted. Please contact your Fisher Scientific Sales Representative for pricing.