Learn More
Beckman Coulter Beckman Coulter Allegra X-30R High Cap Pkg
Supplier: Beckman Coulter B08708
High Capacity Cell Culture Application Package: Allegra X-30R, IVD, 120 V, 60 Hz, Refrigerated Benchtop Centrifuge , 4x 400 mL SX4400 Swinging-bucket Rotor,15 mL Conical Adapters (2 sets of 2), 50 mL Conical Adapters (2 sets of 2). Max. tube capacity: 15 mL = 36 tubes / 50 mL = 16 tubes (Not compatible with optional BioCertified bucket covers.)
This item is not returnable.
View return policy