Learn More
Beckman Coulter Beckman Coulter Allegra X-30R Cell Culture Pkg
Supplier: Beckman Coulter B08540
Cell Culture Application Package: Allegra X-30R, IVD, 120 V, 60 Hz, Refrigerated Benchtop Centrifuge , 4x 400 mL SX4400 Swinging-bucket Rotor,15 mL Conical Adapters (2 sets of 2), 50 mL Conical Adapters (2 sets of 2) Max. tube capacity: 15 mL = 20 tubes / 50 mL = 12 tubes
This item is not returnable.
View return policy