Learn More
Beckman Coulter Beckman Coulter Allegra X-30 Blood Pkg
Supplier: Beckman Coulter B05796
Clinical/Blood Sample Applications Package: Allegra X-30, IVD, 120 V, 60 Hz, Ambient Benchtop Centrifuge, SX4400 (4 x 400mL) Swinging-bucket Rotor, Vacutainer/RIA Adapters/Hemolyse for 10 mL, 16 mm Tubes (2 sets of 2),Vacutainer/Hemolyse/ RIA Adapters for 5 mL, 13 mm Tubes (2 sets of 2)
This item is not returnable.
View return policy