Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meridian Bioscience™ Alethia™ C. difficile Test Kit
Molecular assay for the detection of C. difficiles in stool
Supplier: Meridian Bioscience™ 480050
This item is not returnable.
View return policy
Description
DNA Amplification Assay for the Detection of Cytotoxigenic C. difficile in Stool Specimens.
- Simple, flexible, cost-effective molecular assay for C. difficile detection.
- Allows addition of molecular assays without the cost of additional labor or capital expenditures.
- Specimen Type: Unformed stool
- Assay Format: Isothermal DNA amplification
- CE marked
- Sensitivity: 95.2%
- Specificity: 95.3%
Specifications
C. difficile | |
Alethia Clostridium difficile Test Kit | |
Sample preparation apparatus, dry reagent Lyosphere, C. difficile specific primers, control chamber with S. aureus specific primers | |
50 Tests |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction