Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abbott™ NMP22™ Bladder Cancer ELISA Test Kit
Easy-to-use enzyme immunoassay in 96-well microplate format. Employs two monoclonal antibodies specific for the nuclear matrix protein NMP22.
Supplier: Abbott™ D1100E
This item is not returnable.
View return policy
Description
Provided Materials- Five calibrators, 2mL (lyophilized), ranging in concentration from 0 to ∽120U/mL
- Three tri-level controls, 2mL (lyophilized)
- Coated microplate strip well, 96 wells
- Digoxigenin anti-NMP22 reagent, 20mL
- HRP-SAD reagent, 20mL
- Sample diluent, 10mL (lyophilized)
- Color development buffer, 40mL
- 100x wash concentrate, 30mL
- Four o-phenylenediamine tablets, 10mg
- Note:OPD tablets are known skin and mucous membrane irritants
- 200μL precision pipet with disposable tips
- Precision pipet with disposable tips to deliver 2mL
- Microplate reader (490nm)
- Microplate washer/aspiration device capable of delivering at least 310μL per well of solution with a 10-second soak between aspirations (optional)
- Centrifuge
- Plastic test tubes to prepare OPD solution and specimens
- Two Molar (4 Normal) sulfuric acid stop solution
- Matritech NMP22 Urine Collection Kit (MRD2000, see below)
- Note:Sulfuric acid solution is a known skin and mucous membrane irritant
Specifications
Bladder Cancer | |
NMP22™ Bladder Cancer ELISA Test Kit | |
96 Tests |
Microplate | |
Five calibrators, 2mL (lyophilized), ranging in concentration from 0 to ∼120U/mL, three tri-level control, 2mL (lyophilized), coated microplate strip well, 96 wells, Digoxigenin Anti-NMP22 reagent, 20mL, HRP-SAD reagent, 20mL, sample diluent, 10mL (lyophi |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title