Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abbott™ Flu Starter Kit
Flu Starter Kit combines the DIGIVAL™ Complete Kit (Cat No 23046680) with BinaxNOW™ Influenza A and B Card 2 Test Cards (23046682) and an optional thermal printer (23046687) for all-in-one influenza testing.
Supplier: Abbott™ ARFLUBD
This item is not returnable.
View return policy
Description
Immunochromatographic assay for the qualitative detection of influenza A and B nucleoprotein antigens in nasopharyngeal swabs and nasal swabs.Order Info
Order eligiblity is based upon executed contractual agreement. Please reach out to your Fisher Healthcare Account Representative for more information.
Specifications
Room Temperature | |
Waived | |
Card | |
Lateral Flow Assay | |
Influenza testing | |
Box | |
15 min. |
No | |
Influenza A and B | |
Nasal and NP swabs; Nasal wash/aspirate samples | |
LF test card | |
DIGIVAL Complete Kit; 12 A and B Test Card Sets | |
22 Cards/Set |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction