Learn More
Abbott™ Flu Starter Kit
Flu Starter Kit combines the DIGIVAL™ Complete Kit (Cat No 23046680) with BinaxNOW™ Influenza A and B Card 2 Test Cards (23046682) and an optional thermal printer (23046687) for all-in-one influenza testing.
Supplier: Abbott™ ARFLUPRT
Description
Immunochromatographic assay for the qualitative detection of influenza A and B nucleoprotein antigens in nasopharyngeal swabs and nasal swabs.Order Info
Order eligiblity is based upon executed contractual agreement. Please reach out to your Fisher Healthcare Account Representative for more information.
Specifications
Room Temperature | |
Waived | |
Card | |
Lateral Flow Assay | |
Influenza testing | |
Box | |
15 min. |
No | |
Influenza A and B | |
Nasal and NP swabs; Nasal wash/aspirate samples | |
LF test card | |
DIGIVAL Complete Kit; 3 A and B Test Card Sets; Thermal Printer | |
22 Cards/Set |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.