Learn More
Abbott™ Flu Starter Kit
Flu Starter Kit combines the DIGIVAL™ Complete Kit (Cat No 23046680) with BinaxNOW™ Influenza A and B Card 2 Test Cards (23046682) and an optional thermal printer (23046687) for all-in-one influenza testing.
Supplier:  Abbott™ ARFLUSTART
 View more versions of this product
Catalog No. 23-046-691
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
Immunochromatographic assay for the qualitative detection of influenza A and B nucleoprotein antigens in nasopharyngeal swabs and nasal swabs.
Order Info
Order eligiblity is based upon executed contractual agreement. Please reach out to your Fisher Healthcare Account Representative for more information.
Specifications
Specifications
Room Temperature
Waived
Card
Lateral Flow Assay
Influenza testing
Box
15 min.
No
Influenza A and B
Nasal and NP swabs; Nasal wash/aspirate samples
LF test card
DIGIVAL Complete Kit; 3 A and B Test Card Sets
22 Cards/Set