Learn More
MilliporeSigma™ Supelco™ Aldrich™ TLC Developing Tank - Cylindrical Glass Tank
Aldrich™ TLC developing tanks are used to develop TLC plates.
Supplier: MilliporeSigma™ Supelco™ Z2439143EA
Description
Can be used as a staining chamber. Made of glass with lid. Compact size offers good plate visibility and minimal solvent use.Specifications
Glass | |
TLC | |
21 cm |
3/Pk. | |
Aldrich | |
6.5 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC